siRNA Details
| Gene Symbol: Tnfaip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053382.1 | NP_445834.1 | 4 | 522 | ||
| GenBank mRNA | BC158666.1 | - | 4 | 522 | 8370218 | |
| FQ228771.1 | - | 4 | 584 | |||
| FQ228783.1 | - | 4 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfaip6| Gene Symbol: Tnfaip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053382.1 | NP_445834.1 | 4 | 522 | ||
| GenBank mRNA | BC158666.1 | - | 4 | 522 | 8370218 | |
| FQ228771.1 | - | 4 | 584 | |||
| FQ228783.1 | - | 4 | 584 | |||