siRNA Details
| Gene Symbol: Gnpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053410.1 | NP_445862.1 | 10 | 1458 | ||
| GenBank mRNA | AF218826.1 | - | 10 | 1458 | ||
| FQ217065.1 | - | 1629 | ||||
| FQ223620.1 | - | 1560 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnpat| Gene Symbol: Gnpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053410.1 | NP_445862.1 | 10 | 1458 | ||
| GenBank mRNA | AF218826.1 | - | 10 | 1458 | ||
| FQ217065.1 | - | 1629 | ||||
| FQ223620.1 | - | 1560 | ||||