siRNA Details
| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191711.1 | NP_001178640.1 | 9 | 1134 | ||
| XM_006233099.2 | XP_006233161.1 | 10 | 3640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx20| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191711.1 | NP_001178640.1 | 9 | 1134 | ||
| XM_006233099.2 | XP_006233161.1 | 10 | 3640 | |||