siRNA Details
Gene Symbol: Ddx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191711.1 | NP_001178640.1 | 11 | 1732 | ||
XM_006233099.2 | XP_006233161.1 | 12 | 4238 | |||
GenBank mRNA | AF220455.1 | - | 2 | 478 |
