siRNA Details
Gene Symbol: Ddx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191711.1 | NP_001178640.1 | 4, 5 | 677 | ||
XM_006233099.2 | XP_006233161.1 | 5, 6 | 3183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ddx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191711.1 | NP_001178640.1 | 4, 5 | 677 | ||
XM_006233099.2 | XP_006233161.1 | 5, 6 | 3183 |