siRNA Details
| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053415.1 | NP_445867.1 | 2 | 648 | ||
| GenBank mRNA | AF223642.1 | - | 2 | 648 | ||
| BC091136.1 | - | 1 | 503 | 7376065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcr3| Gene Symbol: Cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053415.1 | NP_445867.1 | 2 | 648 | ||
| GenBank mRNA | AF223642.1 | - | 2 | 648 | ||
| BC091136.1 | - | 1 | 503 | 7376065 | ||