siRNA Details
| Gene Symbol: Fmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053433.2 | NP_445885.2 | 4 | 602 | ||
| GenBank mRNA | AF286595.1 | - | 4 | 446 | ||
| BC087008.1 | - | 4 | 602 | 7129948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo3| Gene Symbol: Fmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053433.2 | NP_445885.2 | 4 | 602 | ||
| GenBank mRNA | AF286595.1 | - | 4 | 446 | ||
| BC087008.1 | - | 4 | 602 | 7129948 | ||