siRNA Details
| Gene Symbol: Stau1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109907.1 | NP_001103377.1 | 10, 11 | 1612 | ||
| NM_053436.2 | NP_445888.1 | 10, 11 | 1630 | |||
| GenBank mRNA | AF227200.1 | - | 10, 11 | 1617 | ||
| AF290989.1 | - | 10, 11 | 1599 | |||
| BC081755.1 | - | 10, 11 | 1570 | |||
| BC101858.1 | - | 10, 11 | 1608 | 7937343 | ||

Stau1
Genomic Map