siRNA Details
| Gene Symbol: Rgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053453.2 | NP_445905.1 | 1 | 38 | ||
| GenBank mRNA | AF279918.1 | - | 51 | |||
| AF321837.1 | - | 1 | 38 | |||
| AJ318489.1 | - | 1 | 38 | |||
| AY043246.1 | - | 62 | ||||
| BC061969.1 | - | 59 | 5621945 | |||
| FQ224353.1 | - | 69 | ||||
| FQ228949.1 | - | 69 | ||||

Rgs2
Genomic Map