siRNA Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235634.1 | XP_006235696.1 | 18 | 3753 | ||
| GenBank mRNA | AF322224.1 | - | 11 | 2577 | ||
| BC088343.1 | - | 1 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa3| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235634.1 | XP_006235696.1 | 18 | 3753 | ||
| GenBank mRNA | AF322224.1 | - | 11 | 2577 | ||
| BC088343.1 | - | 1 | 199 | |||