siRNA Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235634.1 | XP_006235696.1 | 5 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa3| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235634.1 | XP_006235696.1 | 5 | 550 | ||