siRNA Details
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053466.1 | NP_445918.1 | 24, 25 | 3507 | ||
XM_006238452.2 | XP_006238514.1 | 24, 25 | 3680 | |||
XM_006238453.2 | XP_006238515.1 | 24, 25 | 7303 | |||
GenBank mRNA | BC090029.1 | - | 6, 7 | 762 |
