siRNA Details
| Gene Symbol: Sema6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053471.2 | NP_445923.1 | 7, 8 | 893 | ||
| XM_006244348.2 | XP_006244410.1 | 8, 9 | 1362 | |||
| XM_006244349.2 | XP_006244411.1 | 7, 8 | 715 | |||
| GenBank mRNA | AB000776.1 | - | 7, 8 | 893 | ||

Sema6b
Genomic Map