siRNA Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053475.1 | NP_445927.1 | 1 | 486 | ||
| GenBank mRNA | AJ007016.1 | - | 1 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptp4a2| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053475.1 | NP_445927.1 | 1 | 486 | ||
| GenBank mRNA | AJ007016.1 | - | 1 | 486 | ||