siRNA Details
| Gene Symbol: Pola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053479.1 | NP_445931.1 | 19 | 2153 | ||
| GenBank mRNA | AJ011605.1 | - | 19 | 2153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pola1| Gene Symbol: Pola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053479.1 | NP_445931.1 | 19 | 2153 | ||
| GenBank mRNA | AJ011605.1 | - | 19 | 2153 | ||