siRNA Details
| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270802.1 | NP_001257731.1 | 11 | 1701 | ||
| NM_053483.2 | NP_445935.2 | 11 | 1676 | |||
| GenBank mRNA | AJ130946.1 | - | 10 | 1580 | ||
| AY779026.1 | - | 10 | 1570 | |||
| BC062026.1 | - | 11 | 1676 | 5621957 | ||
| BC089787.1 | - | 11 | 1627 | 7326072 | ||
| Gene Symbol: LOC100359600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729596.4 | XP_002729642.1 | 1 | 1696 | ||

Kpna2
Genomic Map