siRNA Details
| Gene Symbol: Cnga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053496.2 | NP_445948.2 | 6 | 1409 | ||
| GenBank mRNA | U12425.1 | - | 6 | 1408 | ||
| U12623.1 | - | 6 | 1410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnga4| Gene Symbol: Cnga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053496.2 | NP_445948.2 | 6 | 1409 | ||
| GenBank mRNA | U12425.1 | - | 6 | 1408 | ||
| U12623.1 | - | 6 | 1410 | |||