siRNA Details
Gene Symbol: Ajuba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053503.1 | NP_445955.1 | 2 | 1443 | ||
GenBank mRNA | AJ306292.1 | - | 2 | 593 | ||
BC085802.1 | - | 2 | 1443 | 7110970 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ajuba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053503.1 | NP_445955.1 | 2 | 1443 | ||
GenBank mRNA | AJ306292.1 | - | 2 | 593 | ||
BC085802.1 | - | 2 | 1443 | 7110970 |