siRNA Details
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053511.1 | NP_445963.1 | 5 | 721 | ||
XM_008764298.1 | XP_008762520.1 | 6 | 924 | |||
GenBank mRNA | AF098301.1 | - | 5 | 721 | ||
FQ214021.1 | - | 5 | 714 |
