siRNA Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053513.1 | NP_445965.1 | 3 | 212 | ||
| XM_008768127.1 | XP_008766349.1 | 2 | 1617 | |||
| XM_008768128.1 | XP_008766350.1 | 3 | 1286 | |||
| XM_008768129.1 | XP_008766351.1 | 3 | 298 | |||
| XM_008768130.1 | XP_008766352.1 | 2 | 143 | |||
| GenBank mRNA | AF087696.1 | - | 2 | 47 | ||
| FQ211962.1 | - | 3 | 397 | |||

Mpp2
Genomic Map