siRNA Details
| Gene Symbol: Mpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053513.1 | NP_445965.1 | 5 | 535 | ||
| XM_008768127.1 | XP_008766349.1 | 4 | 1940 | |||
| XM_008768128.1 | XP_008766350.1 | 5 | 1609 | |||
| XM_008768129.1 | XP_008766351.1 | 5 | 621 | |||
| XM_008768130.1 | XP_008766352.1 | 4 | 466 | |||
| GenBank mRNA | AF087696.1 | - | 4 | 370 | ||
| FQ211962.1 | - | 5 | 720 | |||

Mpp2
Genomic Map