siRNA Details
| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134449.1 | NP_604444.1 | 2 | 458 | ||
| GenBank mRNA | BC101398.1 | - | 2 | 444 | 7933258 | |
| D85435.1 | - | 2 | 458 | |||
| FQ220155.1 | - | 2 | 466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcdbp| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134449.1 | NP_604444.1 | 2 | 458 | ||
| GenBank mRNA | BC101398.1 | - | 2 | 444 | 7933258 | |
| D85435.1 | - | 2 | 458 | |||
| FQ220155.1 | - | 2 | 466 | |||