siRNA Details
| Gene Symbol: Tfpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138870.1 | NP_620225.1 | 1 | 192 | ||
| XM_006228036.2 | XP_006228098.1 | 2 | 219 | |||
| XM_006228037.2 | XP_006228099.1 | 2 | 219 | |||
| GenBank mRNA | AB029495.1 | - | 1 | 192 | ||

Tfpt
Genomic Map