siRNA Details
| Gene Symbol: Slc5a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053521.1 | NP_445973.1 | 2 | 262 | ||
| XM_006244251.2 | XP_006244313.1 | 2 | 365 | |||
| XM_008766774.1 | XP_008764996.1 | 2 | 413 | |||
| XM_008766775.1 | XP_008764997.1 | 2 | 362 | |||
| GenBank mRNA | AB030947.1 | - | 2 | 262 | ||

Slc5a7
Genomic Map