siRNA Details
| Gene Symbol: Abcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109883.2 | NP_001103353.1 | 12, 13 | 1120 | ||
| GenBank mRNA | AF293383.1 | - | 11, 12 | 1016 | ||
| CK356298.1 | - | 6, 7 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcf1| Gene Symbol: Abcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109883.2 | NP_001103353.1 | 12, 13 | 1120 | ||
| GenBank mRNA | AF293383.1 | - | 11, 12 | 1016 | ||
| CK356298.1 | - | 6, 7 | 602 | |||