siRNA Details
| Gene Symbol: Angpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134454.1 | NP_604449.1 | 9 | 1471 | ||
| GenBank mRNA | AF030378.1 | - | 3 | 383 | ||
| AF311728.1 | - | 3 | 383 | |||
| BC161931.1 | - | 9 | 1471 | 9021835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angpt2| Gene Symbol: Angpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134454.1 | NP_604449.1 | 9 | 1471 | ||
| GenBank mRNA | AF030378.1 | - | 3 | 383 | ||
| AF311728.1 | - | 3 | 383 | |||
| BC161931.1 | - | 9 | 1471 | 9021835 | ||