siRNA Details
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053546.1 | NP_445998.1 | 4 | 1023 | ||
GenBank mRNA | AB080023.1 | - | 4 | 1023 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053546.1 | NP_445998.1 | 4 | 1023 | ||
GenBank mRNA | AB080023.1 | - | 4 | 1023 |