siRNA Details
| Gene Symbol: Rfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053547.1 | NP_445999.1 | 2278 | |||
| XM_006250972.2 | XP_006251034.1 | 2340 | ||||
| XM_008770161.1 | XP_008768383.1 | 2343 | ||||
| XM_008770162.1 | XP_008768384.1 | 2340 | ||||
| GenBank mRNA | AF030050.1 | - | 16 | 2198 | ||
| AF059678.1 | - | 2283 | ||||
| CV110819.1 | - | 5 | 522 | |||

Rfc1
Genomic Map