siRNA Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053549.1 | NP_446001.1 | 6, 7 | 788 | ||
| XM_006230911.2 | XP_006230973.1 | 6, 7 | 987 | |||
| GenBank mRNA | BC076395.1 | - | 6, 7 | 788 | 7109119 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vegfb| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053549.1 | NP_446001.1 | 6, 7 | 788 | ||
| XM_006230911.2 | XP_006230973.1 | 6, 7 | 987 | |||
| GenBank mRNA | BC076395.1 | - | 6, 7 | 788 | 7109119 | |