siRNA Details
Gene Symbol: Bud31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053556.1 | NP_446008.1 | 2 | 209 | ||
XM_006248887.1 | XP_006248949.1 | 2 | 326 | |||
XM_006248889.1 | XP_006248951.1 | 3 | 194 | |||
XM_008769003.1 | XP_008767225.1 | 3 | 423 | |||
GenBank mRNA | AF058791.1 | - | 2 | 209 | ||
BC058456.1 | - | 3 | 236 | 6887068 |
