siRNA Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053561.2 | NP_446013.2 | 2, 3 | 184 | ||
XM_006241342.1 | XP_006241404.1 | 2, 3 | 198 | |||
XM_006241343.2 | XP_006241405.1 | 2, 3 | 198 | |||
GenBank mRNA | AF062594.1 | - | 2, 3 | 169 | ||
CO574484.1 | - | 200 |
