siRNA Details
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762869.1 | XP_008761091.1 | 22, 23 | 3441 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762869.1 | XP_008761091.1 | 22, 23 | 3441 |