siRNA Details
| Gene Symbol: Socs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053565.1 | NP_446017.1 | 1 | 508 | ||
| XM_008768398.1 | XP_008766620.1 | 2 | 876 | |||
| GenBank mRNA | AF075383.1 | - | 1 | 508 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs3| Gene Symbol: Socs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053565.1 | NP_446017.1 | 1 | 508 | ||
| XM_008768398.1 | XP_008766620.1 | 2 | 876 | |||
| GenBank mRNA | AF075383.1 | - | 1 | 508 | ||