siRNA Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053589.1 | NP_446041.1 | 5 | 427 | ||
XM_006234049.2 | XP_006234111.1 | 6 | 860 | |||
XM_006234050.2 | XP_006234112.1 | 6 | 512 | |||
GenBank mRNA | BC158585.1 | - | 5 | 1016 | 7446356 | |
M83680.1 | - | 5 | 427 |
