siRNA Details
| Gene Symbol: Nudt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053598.1 | NP_446050.1 | 5 | 578 | ||
| GenBank mRNA | AF253473.1 | - | 5 | 578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt4| Gene Symbol: Nudt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053598.1 | NP_446050.1 | 5 | 578 | ||
| GenBank mRNA | AF253473.1 | - | 5 | 578 | ||