siRNA Details
| Gene Symbol: Nudt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053598.1 | NP_446050.1 | 5 | 606 | ||
| XM_006241282.2 | XP_006241344.1 | 5 | 800 | |||
| GenBank mRNA | AF253473.1 | - | 5 | 606 | ||
| FQ223117.1 | - | 5 | 691 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt4| Gene Symbol: Nudt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053598.1 | NP_446050.1 | 5 | 606 | ||
| XM_006241282.2 | XP_006241344.1 | 5 | 800 | |||
| GenBank mRNA | AF253473.1 | - | 5 | 606 | ||
| FQ223117.1 | - | 5 | 691 | |||