siRNA Details
| Gene Symbol: Kcnj13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053608.2 | NP_446060.1 | 3 | 637 | ||
| XM_008767301.1 | XP_008765523.1 | 2 | 1972 | |||
| GenBank mRNA | AJ006129.1 | - | 2 | 520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj13| Gene Symbol: Kcnj13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053608.2 | NP_446060.1 | 3 | 637 | ||
| XM_008767301.1 | XP_008765523.1 | 2 | 1972 | |||
| GenBank mRNA | AJ006129.1 | - | 2 | 520 | ||