siRNA Details
| Gene Symbol: MAP2K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206804.1 | NP_001193733.1 | 6 | 479 | ||
| NM_002757.3 | NP_002748.1 | 6 | 1063 | |||
| NM_145160.2 | NP_660143.1 | 6 | 1063 | |||
| XM_011521787.2 | XP_011520089.1 | 6 | 1140 | |||
| XM_011521788.2 | XP_011520090.1 | 6 | 1140 | |||
| XM_017022414.1 | XP_016877903.1 | 6 | 1140 | |||
| GenBank mRNA | AK293459.1 | - | 6 | 479 | ||
| AK315435.1 | - | 6 | 610 | |||
| BC008838.2 | - | 6 | 720 | 4100679 | ||
| BT006780.1 | - | 6 | 410 | |||
| CR542229.1 | - | 6 | 410 | |||
| U71087.1 | - | 6 | 471 | |||
| U71088.2 | - | 6 | 415 | |||

MAP2K5
Genomic Map