siRNA Details
| Gene Symbol: MAPK8IP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040439.1 | NP_001035529.1 | 8 | 1236 | ||
| NM_001318852.1 | NP_001305781.1 | 8 | 1276 | |||
| NM_015133.4 | NP_055948.2 | 8 | 1273 | |||
| XM_005255190.2 | XP_005255247.1 | 8 | 1276 | |||
| XM_006720869.2 | XP_006720932.1 | 8 | 2706 | |||
| XM_011522429.2 | XP_011520731.1 | 9 | 1300 | |||
| XM_011522430.2 | XP_011520732.1 | 9 | 1300 | |||
| XM_011522431.2 | XP_011520733.1 | 9 | 1300 | |||
| XM_011522432.2 | XP_011520734.1 | 9 | 1300 | |||
| XM_011522433.2 | XP_011520735.1 | 4 | 1923 | |||
| XM_017023079.1 | XP_016878568.1 | 8 | 1276 | |||
| XM_017023080.1 | XP_016878569.1 | 8 | 1276 | |||
| XM_017023081.1 | XP_016878570.1 | 7 | 748 | |||
| GenBank mRNA | AB028989.2 | - | 8 | 1236 | ||
| BC137123.1 | - | 8 | 1188 | 9021120 | ||
| BC137124.1 | - | 8 | 1188 | 9021121 | ||
| BC144486.1 | - | 8 | 1191 | 9053013 | ||
| BC150266.1 | - | 8 | 1236 | 9007222 | ||
| CA776361.1 | - | 3 | 298 | |||

MAPK8IP3
Genomic Map