siRNA Details
| Gene Symbol: FOXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001455.3 | NP_001446.1 | 2 | 1919 | ||
| NM_201559.2 | NP_963853.1 | 3 | 1892 | |||
| XM_005266867.4 | XP_005266924.1 | 3 | 1140 | |||
| XM_005266868.3 | XP_005266925.1 | 2 | 1164 | |||
| XM_011535626.1 | XP_011533928.1 | 2 | 1090 | |||
| XM_011535628.2 | XP_011533930.1 | 2 | 5448 | |||
| XM_011535629.2 | XP_011533931.1 | 2 | 1113 | |||
| XM_017010585.1 | XP_016866074.1 | 3 | 2825 | |||
| XM_017010586.1 | XP_016866075.1 | 2 | 2708 | |||
| GenBank mRNA | AF032886.1 | - | 2 | 2500 | ||
| AJ001589.1 | - | 2 | 1636 | |||
| AK092357.1 | - | 3 | 1280 | |||
| AK122861.1 | - | 3 | 1475 | |||
| AK301304.1 | - | 2 | 1081 | |||
| AK303933.1 | - | 2 | 1065 | |||
| AK311587.1 | - | 2 | 1179 | |||
| BC020227.1 | - | 3 | 1880 | 4111403 | ||
| BC021224.2 | - | 3 | 1880 | 4137370 | ||
| BC068552.1 | - | 2 | 1919 | 30347471 | ||
| CR749261.1 | - | 2 | 1149 | |||

FOXO3
Genomic Map