siRNA Details
| Gene Symbol: PRKD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005813.4 | NP_005804.1 | 1 | 638 | ||
| XM_005264237.3 | XP_005264294.1 | 2 | 911 | |||
| XM_017003741.1 | XP_016859230.1 | 2 | 861 | |||
| GenBank mRNA | AB015982.2 | - | 1 | 638 | ||
| BC012820.2 | - | 1 | 597 | |||
| BC030706.2 | - | 1 | 583 | 5266769 | ||

PRKD3
Genomic Map