siRNA Details
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027877.1 | 3, 4 | 1360 | |||
GenBank mRNA | AY309002.1 | - | 3, 4 | 1185 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027877.1 | 3, 4 | 1360 | |||
GenBank mRNA | AY309002.1 | - | 3, 4 | 1185 |