siRNA Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027324.1 | 4, 5 | 1728 | |||
| GenBank mRNA | BC099104.1 | - | 4, 5 | 1649 | ||
| BC160921.1 | - | 4, 5 | 1462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H19| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027324.1 | 4, 5 | 1728 | |||
| GenBank mRNA | BC099104.1 | - | 4, 5 | 1649 | ||
| BC160921.1 | - | 4, 5 | 1462 | |||