siRNA Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027324.1 | 1 | 1217 | |||
| GenBank mRNA | BC099104.1 | - | 1 | 1138 | ||
| BC160921.1 | - | 1 | 951 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H19| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027324.1 | 1 | 1217 | |||
| GenBank mRNA | BC099104.1 | - | 1 | 1138 | ||
| BC160921.1 | - | 1 | 951 | |||