siRNA Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027324.1 | 5 | 2309 | |||
| GenBank mRNA | BC099104.1 | - | 5 | 2230 | ||
| BC160921.1 | - | 5 | 2043 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DY319382.1 | - | 631 | Chr. - - [-] on Build | ||

H19
Genomic Map