siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027366.1 | 3 | 1379 | |||
| NR_027367.1 | 3 | 993 | ||||
| GenBank mRNA | BC079219.1 | - | 3 | 1379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhno1| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027366.1 | 3 | 1379 | |||
| NR_027367.1 | 3 | 993 | ||||
| GenBank mRNA | BC079219.1 | - | 3 | 1379 | ||