siRNA Details
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027366.1 | 3 | 1437 | |||
NR_027367.1 | 3 | 1051 | ||||
GenBank mRNA | BC079219.1 | - | 3 | 1437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027366.1 | 3 | 1437 | |||
NR_027367.1 | 3 | 1051 | ||||
GenBank mRNA | BC079219.1 | - | 3 | 1437 |