siRNA Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002704.1 | 10 | 331 | |||
GenBank mRNA | FQ233900.1 | - | 9 | 537 | ||
U77829.1 | - | 10 | 331 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002704.1 | 10 | 331 | |||
GenBank mRNA | FQ233900.1 | - | 9 | 537 | ||
U77829.1 | - | 10 | 331 |