siRNA Details
| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002704.1 | 8, 9 | 280 | |||
| GenBank mRNA | FQ233900.1 | - | 8, 9 | 307 | ||
| U77829.1 | - | 8, 9 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas5| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002704.1 | 8, 9 | 280 | |||
| GenBank mRNA | FQ233900.1 | - | 8, 9 | 307 | ||
| U77829.1 | - | 8, 9 | 280 | |||