siRNA Details
| Gene Symbol: Defb16-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002707.1 | 2 | 133 | |||
| GenBank mRNA | DQ012089.1 | - | 2 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb16-ps| Gene Symbol: Defb16-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002707.1 | 2 | 133 | |||
| GenBank mRNA | DQ012089.1 | - | 2 | 133 | ||